siRNA Details
| Gene Symbol: Kcna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023954.1 | NP_076444.1 | 1 | 1994 | ||
| GenBank mRNA | X17621.1 | - | 1 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna6| Gene Symbol: Kcna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023954.1 | NP_076444.1 | 1 | 1994 | ||
| GenBank mRNA | X17621.1 | - | 1 | 778 | ||