siRNA Details
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022534.1 | NP_071979.1 | 5 | 780 | ||
XM_006251366.2 | XP_006251428.1 | 6 | 971 | |||
XM_006251367.2 | XP_006251429.1 | 6 | 965 | |||
XM_006251368.2 | XP_006251430.1 | 6 | 910 | |||
XM_006251370.2 | XP_006251432.1 | 6 | 903 | |||
GenBank mRNA | AF054810.1 | - | 5 | 780 |
