siRNA Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 4 | 369 | ||
GenBank mRNA | AF071225.1 | - | 4 | 369 | ||
BC061971.1 | - | 4 | 398 | 5623342 | ||
FQ211358.1 | - | 4 | 373 | |||
FQ212031.1 | - | 4 | 398 | |||
FQ219792.1 | - | 4 | 407 | |||
FQ219926.1 | - | 4 | 419 | |||
FQ221020.1 | - | 4 | 422 | |||
FQ221248.1 | - | 4 | 417 | |||
FQ221362.1 | - | 4 | 396 | |||
FQ222696.1 | - | 4 | 531 | |||
FQ223162.1 | - | 4 | 416 | |||
FQ228087.1 | - | 4 | 412 | |||
FQ229174.1 | - | 4 | 419 | |||
FQ229214.1 | - | 4 | 413 | |||
FQ230004.1 | - | 423 | ||||
FQ232077.1 | - | 4 | 408 | |||
FQ232920.1 | - | 4 | 422 |
