siRNA Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031695.2 | NP_113883.2 | 1 | 89 | ||
| XM_006255625.2 | XP_006255687.1 | 3 | 1264 | |||
| XM_006255626.2 | XP_006255688.1 | 4 | 1456 | |||
| XM_006255627.2 | XP_006255689.1 | 2 | 1146 | |||
| GenBank mRNA | X76988.1 | - | 101 | |||

St3gal2
Genomic Map