siRNA Details
| Gene Symbol: Cited1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172055.2 | NP_742052.2 | 3 | 598 | ||
| XM_006257097.2 | XP_006257159.1 | 3 | 723 | |||
| XM_006257098.2 | XP_006257160.1 | 4 | 1287 | |||
| XM_006257099.2 | XP_006257161.1 | 3 | 1169 | |||
| XM_006257100.2 | XP_006257162.1 | 3 | 613 | |||
| GenBank mRNA | AF104399.1 | - | 2 | 456 | ||
| BC097276.1 | - | 3 | 598 | 7446633 | ||

Cited1
Genomic Map