siRNA Details
| Gene Symbol: Entpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172030.1 | NP_742027.1 | 6 | 1014 | ||
| GenBank mRNA | AF276940.1 | - | 6 | 1014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd2| Gene Symbol: Entpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172030.1 | NP_742027.1 | 6 | 1014 | ||
| GenBank mRNA | AF276940.1 | - | 6 | 1014 | ||