siRNA Details
| Gene Symbol: Slc30a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172066.1 | NP_742063.1 | 7 | 1531 | ||
| GenBank mRNA | BC086981.1 | - | 7 | 1497 | 7112403 | |
| Y16774.1 | - | 7 | 1531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a4| Gene Symbol: Slc30a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172066.1 | NP_742063.1 | 7 | 1531 | ||
| GenBank mRNA | BC086981.1 | - | 7 | 1497 | 7112403 | |
| Y16774.1 | - | 7 | 1531 | |||