siRNA Details
| Gene Symbol: Hdlbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172039.2 | NP_742036.2 | 11, 12 | 1540 | ||
| XM_006245548.1 | XP_006245610.1 | 11, 12 | 1580 | |||
| GenBank mRNA | BC105748.1 | - | 11, 12 | 1540 | 5598374 | |
| U90725.1 | - | 11, 12 | 1540 | |||

Hdlbp
Genomic Map