siRNA Details
| Gene Symbol: Pim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022602.1 | NP_072124.1 | 1079 | |||
| GenBank mRNA | AF057026.1 | - | 1079 | |||
| AF086624.1 | - | 1281 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pim3| Gene Symbol: Pim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022602.1 | NP_072124.1 | 1079 | |||
| GenBank mRNA | AF057026.1 | - | 1079 | |||
| AF086624.1 | - | 1281 | ||||