siRNA Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022607.2 | NP_072129.2 | 3 | 880 | ||
| GenBank mRNA | AB000098.1 | - | 3 | 811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufv3| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022607.2 | NP_072129.2 | 3 | 880 | ||
| GenBank mRNA | AB000098.1 | - | 3 | 811 | ||