siRNA Details
| Gene Symbol: Bcl2l11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022612.1 | NP_072134.1 | 3, 4 | 325 | ||
| NM_171988.2 | NP_741985.1 | 3, 4 | 756 | |||
| NM_171989.1 | NP_741986.1 | 2, 3 | 235 | |||
| XM_006234983.2 | XP_006235045.2 | 3, 4 | 874 | |||
| XM_006234985.2 | XP_006235047.2 | 4, 5 | 706 | |||
| XM_006234987.2 | XP_006235049.2 | 3, 4 | 616 | |||
| GenBank mRNA | AF065431.1 | - | 2, 3 | 184 | ||
| AF065432.1 | - | 2, 3 | 235 | |||
| AF065433.1 | - | 2, 3 | 493 | |||
| AF136927.1 | - | 3, 4 | 325 | |||

Bcl2l11
Genomic Map