siRNA Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022615.1 | NP_072137.1 | 14 | 1394 | ||
| GenBank mRNA | AF140782.1 | - | 14 | 1394 | ||
| Gene Symbol: LOC499179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759598.1 | XP_008757820.1 | 4 | 1532 | ||

Top1
Genomic Map