siRNA Details
| Gene Symbol: Prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022627.2 | NP_072149.1 | 4 | 527 | ||
| GenBank mRNA | AF182717.1 | - | 4 | 529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkab2| Gene Symbol: Prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022627.2 | NP_072149.1 | 4 | 527 | ||
| GenBank mRNA | AF182717.1 | - | 4 | 529 | ||