siRNA Details
Gene Symbol: Vax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022637.1 | NP_072159.1 | 3 | 543 | ||
GenBank mRNA | AF113516.1 | - | 3 | 543 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022637.1 | NP_072159.1 | 3 | 543 | ||
GenBank mRNA | AF113516.1 | - | 3 | 543 |