siRNA Details
| Gene Symbol: Hes3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022687.1 | NP_073178.1 | 3 | 404 | ||
| XM_008764381.1 | XP_008762603.1 | 4 | 501 | |||
| XM_008764382.1 | XP_008762604.1 | 4 | 437 | |||
| XM_008764383.1 | XP_008762605.1 | 4 | 379 | |||
| GenBank mRNA | D13418.1 | - | 3 | 404 | ||

Hes3
Genomic Map