siRNA Details
| Gene Symbol: Hes3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022687.1 | NP_073178.1 | 3, 4 | 436 | ||
| XM_008764381.1 | XP_008762603.1 | 4, 5 | 533 | |||
| XM_008764382.1 | XP_008762604.1 | 4, 5 | 469 | |||
| XM_008764383.1 | XP_008762605.1 | 4, 5 | 411 | |||
| GenBank mRNA | D13418.1 | - | 3, 4 | 436 | ||

Hes3
Genomic Map