siRNA Details
| Gene Symbol: Hand2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022696.1 | NP_073187.1 | 485 | |||
| GenBank mRNA | Y08138.1 | - | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hand2| Gene Symbol: Hand2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022696.1 | NP_073187.1 | 485 | |||
| GenBank mRNA | Y08138.1 | - | 485 | |||