siRNA Details
| Gene Symbol: Hist3h2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021840.2 | NP_068612.2 | 1 | 32 | ||
| GenBank mRNA | BC099140.1 | - | 1 | 32 | 7457173 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist3h2a| Gene Symbol: Hist3h2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021840.2 | NP_068612.2 | 1 | 32 | ||
| GenBank mRNA | BC099140.1 | - | 1 | 32 | 7457173 | |