siRNA Details
| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031698.1 | NP_113886.1 | 12 | 1377 | ||
| GenBank mRNA | X55298.1 | - | 12 | 1377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpn2| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031698.1 | NP_113886.1 | 12 | 1377 | ||
| GenBank mRNA | X55298.1 | - | 12 | 1377 | ||