siRNA Details
| Gene Symbol: Nolc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022869.1 | NP_074060.1 | 8, 9 | 1067 | ||
| XM_006231560.2 | XP_006231622.1 | 8, 9 | 1115 | |||
| GenBank mRNA | M94287.1 | - | 8, 9 | 1067 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nolc1| Gene Symbol: Nolc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022869.1 | NP_074060.1 | 8, 9 | 1067 | ||
| XM_006231560.2 | XP_006231622.1 | 8, 9 | 1115 | |||
| GenBank mRNA | M94287.1 | - | 8, 9 | 1067 | ||