siRNA Details
| Gene Symbol: Rwdd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128152.1 | NP_001121624.1 | 4 | 768 | ||
| XM_008761472.1 | XP_008759694.1 | 3 | 848 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rwdd3| Gene Symbol: Rwdd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128152.1 | NP_001121624.1 | 4 | 768 | ||
| XM_008761472.1 | XP_008759694.1 | 3 | 848 | |||