siRNA Details
| Gene Symbol: Fat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022954.1 | NP_075243.1 | 2 | 676 | ||
| GenBank mRNA | AB011527.1 | - | 2 | 676 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat2| Gene Symbol: Fat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022954.1 | NP_075243.1 | 2 | 676 | ||
| GenBank mRNA | AB011527.1 | - | 2 | 676 | ||