siRNA Details
| Gene Symbol: Cldn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031700.2 | NP_113888.2 | 1 | 752 | ||
| GenBank mRNA | BC062411.1 | - | 1 | 377 | 5623891 | |
| M74067.1 | - | 1 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn3| Gene Symbol: Cldn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031700.2 | NP_113888.2 | 1 | 752 | ||
| GenBank mRNA | BC062411.1 | - | 1 | 377 | 5623891 | |
| M74067.1 | - | 1 | 472 | |||