siRNA Details
Gene Symbol: Cldn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031700.2 | NP_113888.2 | 1 | 795 | ||
GenBank mRNA | BC062411.1 | - | 1 | 420 | 5623891 | |
M74067.1 | - | 1 | 515 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cldn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031700.2 | NP_113888.2 | 1 | 795 | ||
GenBank mRNA | BC062411.1 | - | 1 | 420 | 5623891 | |
M74067.1 | - | 1 | 515 |