siRNA Details
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031715.1 | NP_113903.1 | 1431 | |||
GenBank mRNA | BC094212.1 | - | 14 | 1418 | 7314667 | |
FN433010.1 | - | 14 | 1396 | |||
FQ211719.1 | - | 1557 | ||||
U25651.1 | - | 1431 |
