siRNA Details
| Gene Symbol: Pfkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031715.1 | NP_113903.1 | 875 | |||
| GenBank mRNA | BC094212.1 | - | 8, 9 | 862 | 7314667 | |
| FN433010.1 | - | 8, 9 | 840 | |||
| FQ211719.1 | - | 1001 | ||||
| U25651.1 | - | 875 | ||||

Pfkm
Genomic Map