siRNA Details
| Gene Symbol: Ncs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024366.1 | NP_077342.1 | 4, 5 | 368 | ||
| XM_006233925.2 | XP_006233987.1 | 4, 5 | 672 | |||
| XM_008761679.1 | XP_008759901.1 | 3, 4 | 263 | |||
| GenBank mRNA | L27421.1 | - | 4, 5 | 308 | ||
| X82188.1 | - | 4, 5 | 368 | |||

Ncs1
Genomic Map