siRNA Details
| Gene Symbol: Amotl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031717.1 | NP_113905.1 | 2 | 457 | ||
| XM_006243680.2 | XP_006243742.1 | 2 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amotl2| Gene Symbol: Amotl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031717.1 | NP_113905.1 | 2 | 457 | ||
| XM_006243680.2 | XP_006243742.1 | 2 | 559 | |||