siRNA Details
| Gene Symbol: Litaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105735.2 | NP_001099205.1 | 1 | 49 | ||
| GenBank mRNA | BC166851.1 | - | 1 | 49 | 9091522 | |
| U53184.1 | - | 86 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Litaf| Gene Symbol: Litaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105735.2 | NP_001099205.1 | 1 | 49 | ||
| GenBank mRNA | BC166851.1 | - | 1 | 49 | 9091522 | |
| U53184.1 | - | 86 | ||||