siRNA Details
| Gene Symbol: Nkx6-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031737.1 | NP_113925.1 | 3 | 1838 | ||
| GenBank mRNA | AF004431.1 | - | 3 | 1838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx6-1| Gene Symbol: Nkx6-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031737.1 | NP_113925.1 | 3 | 1838 | ||
| GenBank mRNA | AF004431.1 | - | 3 | 1838 | ||