siRNA Details
Gene Symbol: Kcnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270962.1 | NP_001257891.1 | 2 | 635 | ||
NM_001270963.1 | NP_001257892.1 | 2 | 635 | |||
NM_031739.3 | NP_113927.2 | 2 | 635 | |||
GenBank mRNA | AB003587.1 | - | 1 | 543 | ||
AF334791.1 | - | 1 | 543 | |||
L48619.1 | - | 2 | 642 | |||
M74898.1 | - | 1 | 36 | |||
U75448.1 | - | 1 | 563 |
