siRNA Details
| Gene Symbol: Trnau1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023027.1 | NP_075416.1 | 3 | 141 | ||
| GenBank mRNA | AF181856.1 | - | 3 | 141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trnau1ap| Gene Symbol: Trnau1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023027.1 | NP_075416.1 | 3 | 141 | ||
| GenBank mRNA | AF181856.1 | - | 3 | 141 | ||