siRNA Details
Gene Symbol: Gpr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023099.1 | NP_075587.1 | 1 | 918 | ||
GenBank mRNA | AB040802.1 | - | 1 | 918 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
