siRNA Details
| Gene Symbol: Rhox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037581.1 | NP_001032670.1 | 1 | 343 | ||
| GenBank mRNA | DQ058657.1 | - | 1 | 343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox10| Gene Symbol: Rhox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037581.1 | NP_001032670.1 | 1 | 343 | ||
| GenBank mRNA | DQ058657.1 | - | 1 | 343 | ||