siRNA Details
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100804.1 | NP_001094274.1 | 1 | 44 | ||
GenBank mRNA | AY262357.1 | - | 1 | 33 | ||
BC168196.1 | - | 44 | 7111460 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100804.1 | NP_001094274.1 | 1 | 44 | ||
GenBank mRNA | AY262357.1 | - | 1 | 33 | ||
BC168196.1 | - | 44 | 7111460 |