siRNA Details
Gene Symbol: Rnf130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037658.1 | NP_001032747.1 | 1264 | |||
GenBank mRNA | AY190520.1 | - | 1264 | |||
BC166595.1 | - | 1429 | 5599048 | |||
FQ234228.1 | - | 5 | 754 |
