siRNA Details
| Gene Symbol: LOC652956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037659.1 | NP_001032748.1 | 938 | |||
| GenBank mRNA | AB098073.1 | - | 938 | |||
| BC091223.1 | - | 834 | 7127032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC652956| Gene Symbol: LOC652956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037659.1 | NP_001032748.1 | 938 | |||
| GenBank mRNA | AB098073.1 | - | 938 | |||
| BC091223.1 | - | 834 | 7127032 | |||