siRNA Details
| Gene Symbol: Spag11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037850.1 | NP_001032939.1 | 2 | 168 | ||
| NM_001037852.1 | NP_001032941.1 | 2 | 228 | |||
| GenBank mRNA | DQ012092.1 | - | 2 | 168 | ||
| DQ012093.1 | - | 2 | 228 | |||
| Gene Symbol: Spag11bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728419.4 | XP_002728465.2 | 2 | 355 | ||
| XM_006253377.2 | XP_006253439.1 | 2 | 358 | |||
| GenBank mRNA | AY600144.1 | - | 2 | 168 | ||
| AY600145.1 | - | 2 | 188 | |||

Spag11b
Genomic Map