siRNA Details
| Gene Symbol: Cox6b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039085.1 | NP_001034174.1 | 2 | 172 | ||
| XM_008758979.1 | XP_008757201.1 | 2 | 305 | |||
| GenBank mRNA | AY152399.1 | - | 2 | 172 | ||
| CK840972.1 | - | 3 | 313 | |||

Cox6b2
Genomic Map