siRNA Details
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039346.1 | NP_001034435.1 | 9 | 1154 | ||
GenBank mRNA | AY886532.1 | - | 9 | 1154 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039346.1 | NP_001034435.1 | 9 | 1154 | ||
GenBank mRNA | AY886532.1 | - | 9 | 1154 |