siRNA Details
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039346.1 | NP_001034435.1 | 3 | 435 | ||
XM_006231403.2 | XP_006231465.1 | 3 | 471 | |||
XM_006231405.2 | XP_006231467.1 | 3 | 472 | |||
XM_006231407.2 | XP_006231469.1 | 3 | 472 | |||
GenBank mRNA | AY886532.1 | - | 3 | 435 | ||
BC127439.1 | - | 3 | 436 | 8367792 |
