siRNA Details
| Gene Symbol: Avgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039588.1 | NP_001034677.1 | 709 | |||
| GenBank mRNA | DQ366675.1 | - | 709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avgr1| Gene Symbol: Avgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039588.1 | NP_001034677.1 | 709 | |||
| GenBank mRNA | DQ366675.1 | - | 709 | |||