siRNA Details
| Gene Symbol: Fgf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023961.1 | NP_076451.1 | 3 | 482 | ||
| GenBank mRNA | AB020021.1 | - | 3 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf20| Gene Symbol: Fgf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023961.1 | NP_076451.1 | 3 | 482 | ||
| GenBank mRNA | AB020021.1 | - | 3 | 482 | ||