siRNA Details
| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023967.1 | NP_076457.1 | 5 | 767 | ||
| XM_006235040.2 | XP_006235102.1 | 3 | 2981 | |||
| XM_006235041.2 | XP_006235103.1 | 4 | 2240 | |||
| XM_006235042.2 | XP_006235104.1 | 4 | 2240 | |||
| XM_006235043.2 | XP_006235105.1 | 4 | 2240 | |||
| XM_006235044.2 | XP_006235106.1 | 3 | 2105 | |||
| XM_006235045.2 | XP_006235107.1 | 5 | 1680 | |||
| XM_008762196.1 | XP_008760418.1 | 4 | 2421 | |||
| XM_008762197.1 | XP_008760419.1 | 5 | 1680 | |||
| GenBank mRNA | AJ294475.1 | - | 5 | 767 | ||
| AJ294476.1 | - | 5 | 767 | |||

Gfra4
Genomic Map