siRNA Details
| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023967.1 | NP_076457.1 | 5, 6 | 795 | ||
| XM_006235043.2 | XP_006235105.1 | 4, 5 | 2268 | |||
| GenBank mRNA | AJ294476.1 | - | 5, 6 | 795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfra4| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023967.1 | NP_076457.1 | 5, 6 | 795 | ||
| XM_006235043.2 | XP_006235105.1 | 4, 5 | 2268 | |||
| GenBank mRNA | AJ294476.1 | - | 5, 6 | 795 | ||