siRNA Details
| Gene Symbol: Lpar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023969.1 | NP_076459.1 | 1 | 337 | ||
| XM_006233469.2 | XP_006233531.1 | 1 | 377 | |||
| XM_006233470.2 | XP_006233532.1 | 2 | 545 | |||
| XM_006233471.2 | XP_006233533.1 | 1 | 378 | |||
| XM_008761522.1 | XP_008759744.1 | 2 | 592 | |||
| XM_008761523.1 | XP_008759745.1 | 2 | 1311 | |||
| GenBank mRNA | AB051164.1 | - | 1 | 365 | ||
| AF097733.1 | - | 1 | 337 | |||
More Information
Set Membership: |
|

Lpar3
Genomic Map