siRNA Details
| Gene Symbol: Teddm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039915.2 | NP_001035004.1 | 1 | 81 | ||
| GenBank mRNA | AJ515382.1 | - | 1 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Teddm1| Gene Symbol: Teddm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039915.2 | NP_001035004.1 | 1 | 81 | ||
| GenBank mRNA | AJ515382.1 | - | 1 | 82 | ||