siRNA Details
Gene Symbol: Mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059437.5 | XP_001059437.3 | 2 | 351 | ||
GenBank mRNA | BC158892.1 | - | 2 | 172 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059437.5 | XP_001059437.3 | 2 | 351 | ||
GenBank mRNA | BC158892.1 | - | 2 | 172 |