siRNA Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001059437.5 | XP_001059437.3 | 2 | 259 | ||
| GenBank mRNA | BC158892.1 | - | 2 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd4| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001059437.5 | XP_001059437.3 | 2 | 259 | ||
| GenBank mRNA | BC158892.1 | - | 2 | 80 | ||