siRNA Details
| Gene Symbol: LOC679149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100999.1 | NP_001094469.1 | 77 | |||
| GenBank mRNA | BC097486.1 | - | 77 | 7459735 | ||
| CV119728.1 | - | 1 | 76 | |||
| Gene Symbol: Ces2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271303.1 | NP_001258232.1 | 1 | 98 | ||
| Gene Symbol: LOC100365112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190369.1 | NP_001177298.1 | 1 | 100 | ||
| GenBank mRNA | AB010632.1 | - | 1 | 70 | ||

LOC679149
Genomic Map